Benzylidenaceton, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Phenylbut-3-en-2-one, 4-Phenyl-3-buten-2-one
M-Clarity™ Qualitätsstufe = MQ100
147885-250G
SIAL147885-250GEA
51.8
EUR
SIAL147885-250G
Benzylidenaceton, Sigma-Aldrich®
Benzylidenaceton
Formel:
C₁₀H₁₀O Molecular Weight: 146,19 g/mol |
MDL:
MFCD00008779 CAS-Nummer: 122-57-6 EINECS: 204-555-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...