1,8-Bis(dimethylamino)naphthalen, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N1,N1,N8,N8-Tetramethylnaphthalene-1,8-diamine

M-Clarity™ Qualitätsstufe = MQ200

14795-10G 14795-50G
SIAL14795-10GEA 55.1 EUR
SIAL14795-10G SIAL14795-50G
1,8-Bis(dimethylamino)naphthalen, Sigma-Aldrich®
1,8-Bis(dimethylamino)naphthalen
Formel: C₁₄H₁₈N₂
Molecular Weight: 214.31 g/mol
MDL: MFCD00003920
CAS-Nummer: 20734-58-1
EINECS: 244-001-6
UN: 3263
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR