Hexaphenylbenzol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3',4',5',6'-Tetraphenyl-o-terphenyl
M-Clarity™ Qualitätsstufe = MQ100
149454-1G
SIAL149454-1GEA
50.3
EUR
SIAL149454-1G
Hexaphenylbenzol, Sigma-Aldrich®
Hexaphenylbenzol
Formel:
C₄₂H₃₀ Molecular Weight: 534,7 g/mol Schmelzpunkt: > 300 °C |
MDL:
MFCD00003057 CAS-Nummer: 992-04-1 EINECS: 213-591-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...