Fluoren-2-carbaldehyd, Sigma-Aldrich®

Synonyme: Fluorene-2-carboxaldehyde

M-Clarity™ Qualitätsstufe = MQ100

150142-5G
SIAL150142-5GEA 160 EUR
SIAL150142-5G
Fluoren-2-carbaldehyd, Sigma-Aldrich®
Fluoren-2-carbaldehyd
Formel: C₁₄H₁₀O
Molecular Weight: 194.23 g/mol
MDL: MFCD00001123
CAS-Nummer: 30084-90-3
EINECS: 250-035-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR