5-Bromcytosin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-5-brom-2-pyrimidinol, 4-Amino-5-brom-2-hydroxypyrimidin

M-Clarity™ Qualitätsstufe = MQ100

150649-1G
SIAL150649-1GEA 168 EUR
SIAL150649-1G
5-Bromcytosin, Sigma-Aldrich®
5-Bromcytosin
Formel: C₄H₄BrN₃O
Molecular Weight: 190 g/mol
Schmelzpunkt: 240…243 °C
MDL: MFCD00056025
CAS-Nummer: 2240-25-7
EINECS: 218-806-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR