Cholesterololeylcarbonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (3S,8S,9S,10R,13R,14S,17R)-10,13-Dimethyl-17-((R)-6-methylheptan-2-yl)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl (Z)-octadec-9-en-1-yl carbonate

M-Clarity™ Qualitätsstufe = MQ100

151157-25G
SIAL151157-25GEA 108 EUR
SIAL151157-25G
Cholesterololeylcarbonat, Sigma-Aldrich®
Cholesterololeylcarbonat
Formel: C₄₆H₈₀O₃
Molecular Weight: 681,125 g/mol
MDL: MFCD00003634
CAS-Nummer: 17110-51-9
EINECS: 241-179-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR