N(α)-Boc-D-histidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Boc-D-His-OH

15119-1G
SIAL15119-1GEA 128 EUR
SIAL15119-1G
N(α)-Boc-D-histidin, Sigma-Aldrich®
N(α)-Boc-D-histidin
Formel: C₁₁H₁₇N₃O₄
Molecular Weight: 255.3 g/mol
MDL: MFCD00037851
CAS-Nummer: 50654-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR