DL-3,4-Dihydroxymandelsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(±)-2-(3,4-Dihydroxyphenyl)-2-hydroxyessigsäure, (±)-3,4-Dihydroxyphenylglykolsäure, (±)-2-(3,4-Dihydroxyphenyl)-2-hydroxyethansäure
M-Clarity™ Qualitätsstufe = MQ100
151610-500MG
SIAL151610-500MGEA
216
EUR
SIAL151610-500MG
DL-3,4-Dihydroxymandelsäure, Sigma-Aldrich®
DL-3,4-Dihydroxymandelsäure
Formel:
C₈H₈O₅ Molecular Weight: 184,15 g/mol Schmelzpunkt: 133…137 °C |
MDL:
MFCD00004231 CAS-Nummer: 14883-87-5 EINECS: 238-956-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...