N(α)-Boc-D-tryptophan, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Boc-D-Trp-OH

15185-5G
SIAL15185-5GEA 78.7 EUR
SIAL15185-5G
N(α)-Boc-D-tryptophan, Sigma-Aldrich®
N(α)-Boc-D-tryptophan
Formel: C₁₆H₂₀N₂O₄
Molecular Weight: 304,3 g/mol
MDL: MFCD00037944
CAS-Nummer: 5241-64-5
EINECS: 226-042-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR