Phenoxyaceton, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Phenoxypropan-2-one
152021-5G
SIAL152021-5GEA
87.5
EUR
SIAL152021-5G
Phenoxyaceton, Sigma-Aldrich®
Phenoxyaceton
Formel:
(C₆H₅)OCH₂C(O)CH₃ Molecular Weight: 150,18 g/mol Siedepunkt: 45…47 °C (0,1 mmHg) Dichte: 1,0852 g/cm³ (20 °C) |
MDL:
MFCD00008767 CAS-Nummer: 621-87-4 EINECS: 210-712-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...