4-Methoxyphenylhydraziniumchlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

152137-10G 152137-50G
SIAL152137-10GEA 120 EUR
SIAL152137-10G SIAL152137-50G
4-Methoxyphenylhydraziniumchlorid, Sigma-Aldrich®
4-Methoxyphenylhydraziniumchlorid
Formel: C₇H₁₁ClN₂O
MDL: MFCD00012945
CAS-Nummer: 19501-58-7
EINECS: 243-115-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR