Bistrifluoracetamid, Sigma-Aldrich®

Lieferant: Merck

15225-5G-F
SIAL15225-5G-FEA 265 EUR
SIAL15225-5G-F
Bistrifluoracetamid, Sigma-Aldrich®
Bistrifluoracetamid
Formel: C₄HF₆NO₂
MDL: MFCD00013564
CAS-Nummer: 407-24-9
EINECS: 206-981-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR