1,3-Bis(trimethylsilyl)harnstoff, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N,N'-Bis(trimethylsilyl)urea

M-Clarity™ Qualitätsstufe = MQ100

15248-250G
SIAL15248-250GEA 183 EUR
SIAL15248-250G
1,3-Bis(trimethylsilyl)harnstoff, Sigma-Aldrich®
1,3-Bis(trimethylsilyl)harnstoff
Formel: C₇H₂₀N₂OSi₂
Molecular Weight: 204,42 g/mol
MDL: MFCD00008260
CAS-Nummer: 18297-63-7
EINECS: 242-177-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR