5-Brom-2-thiophencarbaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

152625-100G 152625-25G
SIAL152625-100GEA 541 EUR
SIAL152625-100G SIAL152625-25G
5-Brom-2-thiophencarbaldehyd, Sigma-Aldrich®
5-Brom-2-thiophencarbaldehyd
Formel: C₅H₃BrOS
MDL: MFCD00005432
CAS-Nummer: 4701-17-1
EINECS: 225-176-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR