Bis(trimethylsilylmethyl)sulfid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Bis(trimethylsilylmethyl) sulphide
15264-5ML-F
SIAL15264-5ML-FEA
166
EUR
SIAL15264-5ML-F
Bis(trimethylsilylmethyl)sulfid, Sigma-Aldrich®
Bis(trimethylsilylmethyl)sulfid
Formel:
C₈H₂₂SSi₂ Molecular Weight: 206,5 g/mol |
MDL:
MFCD00039797 CAS-Nummer: 4712-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...