2,4-Dichlorphenylhydraziniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(2,4-Dichlorphenyl)hydrazin Hydrochlorid, 2,4-Dichlorphenylhydrazinhydrochlorid, 2,4-Dichlorphenylhydrazin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

152803-5G
SIAL152803-5GEA 26.2 EUR
SIAL152803-5G
2,4-Dichlorphenylhydraziniumchlorid, Sigma-Aldrich®
2,4-Dichlorphenylhydraziniumchlorid
Formel: C₆H₇Cl₃N₂
Molecular Weight: 213,49 g/mol
MDL: MFCD00012929
CAS-Nummer: 5446-18-4
EINECS: 226-659-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR