3,4-Dichlorphenylhydraziniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,4-Dichlorphenylhydrazinhydrochlorid, 1-(3,4-Dichlorphenyl)hydrazin Hydrochlorid, 3,4-Dichlorphenylhydrazin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

152811-25G
SIAL152811-25GEA 175 EUR
SIAL152811-25G
3,4-Dichlorphenylhydraziniumchlorid, Sigma-Aldrich®
3,4-Dichlorphenylhydraziniumchlorid
Formel: C₆H₇Cl₃N₂
Molecular Weight: 213.49 g/mol
MDL: MFCD00012937
CAS-Nummer: 19763-90-7
EINECS: 243-278-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR