1-Benzyl-4-hydroxypiperidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Benzylpiperidin-4-ol

152986-25G
SIAL152986-25GEA 79.3 EUR
SIAL152986-25G
1-Benzyl-4-hydroxypiperidin, Sigma-Aldrich®
1-Benzyl-4-hydroxypiperidin
Formel: C₁₂H₁₇NO
Molecular Weight: 191,27 g/mol
MDL: MFCD00006503
CAS-Nummer: 4727-72-4
EINECS: 225-226-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR