2'-Aminobenzophenon-2-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2'-Aminobenzophenone-2-carboxylic Acid, 2-Anthraniloylbenzoic acid, 2-(2-Aminobenzoyl)benzoic acid

M-Clarity™ Qualitätsstufe = MQ100

153273-1G
SIAL153273-1GEA 57.8 EUR
SIAL153273-1G
2'-Aminobenzophenon-2-carbonsäure, Sigma-Aldrich®
2'-Aminobenzophenon-2-carbonsäure
Formel: C₁₄H₁₁NO₃
Molecular Weight: 241,2420 g/mol
MDL: MFCD00007715
CAS-Nummer: 1147-43-9
EINECS: 214-554-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR