2,6-Dichlorphenylhydraziniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Dichlorphenylhydrazinhydrochlorid, 2,6-Dichlorphenylhydrazin Hydrochlorid, 1-(2,6-Dichlorphenyl)hydrazin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

153419-1G
SIAL153419-1GEA 54.5 EUR
SIAL153419-1G
2,6-Dichlorphenylhydraziniumchlorid, Sigma-Aldrich®
2,6-Dichlorphenylhydraziniumchlorid
Formel: C₆H₇Cl₃N₂
Molecular Weight: 213.49 g/mol
MDL: MFCD00012930
CAS-Nummer: 50709-36-9
EINECS: 256-730-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR