2-Fluorphenylhydraziniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Fluorphenylhydrazin Hydrochlorid, 1-(2-Fluorphenyl)hydrazin Hydrochlorid, 2-Fluorphenylhydrazinhydrochlorid

M-Clarity™ Qualitätsstufe = MQ200

153427-25G
SIAL153427-25GEA 80.4 EUR
SIAL153427-25G
2-Fluorphenylhydraziniumchlorid, Sigma-Aldrich®
2-Fluorphenylhydraziniumchlorid
Formel: C₆H₈ClFN₂
Molecular Weight: 162.59 g/mol
MDL: MFCD00012927
CAS-Nummer: 2924-15-4
EINECS: 220-885-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR