2-Cyan-4-nitroanilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Nitroanthranilonitril, 2-Amino-5-nitrobenzoesäurenitril, 2-Amino-5-nitrobenzonitril

M-Clarity™ Qualitätsstufe = MQ100

153494-100G
SIAL153494-100GEA 36 EUR
SIAL153494-100G
2-Cyan-4-nitroanilin, Sigma-Aldrich®
2-Cyan-4-nitroanilin
Formel: C₇H₅N₃O₂
Molecular Weight: 163,14 g/mol
Schmelzpunkt: 210 °C
MDL: MFCD00007362
CAS-Nummer: 17420-30-3
EINECS: 241-446-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR