2,4,6-Triphenoxy-1,3,5-triazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,4,6-Triphenoxy-s-triazin

M-Clarity™ Qualitätsstufe = MQ100

153664-10G
SIAL153664-10GEA 35.4 EUR
SIAL153664-10G
2,4,6-Triphenoxy-1,3,5-triazin, Sigma-Aldrich®
2,4,6-Triphenoxy-1,3,5-triazine
Formel: C₂₁H₁₅N₃O₃
Molecular Weight: 357,37 g/mol
MDL: MFCD00006048
CAS-Nummer: 1919-48-8
EINECS: 217-644-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR