(±)-4-Brom-α-methylbenzylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Bromo-α-methylbenzylamine, (+/-)-1-(4-Bromophenyl)ethylamine, (±)-4-Bromo-α-methylbenzylamine

M-Clarity™ Qualitätsstufe = MQ200

154067-10G
SIAL154067-10GEA 66.5 EUR
SIAL154067-10G
(±)-4-Brom-α-methylbenzylamin, Sigma-Aldrich®
(±)-4-Brom-α-methylbenzylamin
Formel: C₈H₁₀BrN
Molecular Weight: 200,08 g/mol
MDL: MFCD00025548
CAS-Nummer: 24358-62-1
EINECS: 246-200-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR