2-(4-Methoxyphenyl)ethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(4-Methoxyphenyl)ethylalkohol, 4-Methoxyphenethylalkohol

M-Clarity™ Qualitätsstufe = MQ100

154180-10G
SIAL154180-10GEA 43.4 EUR
SIAL154180-10G
2-(4-Methoxyphenyl)ethanol, Sigma-Aldrich®
2-(4-Methoxyphenyl)ethanol
Formel: C₉H₁₂O₂
Molecular Weight: 152,19 g/mol
MDL: MFCD00002900
CAS-Nummer: 702-23-8
EINECS: 211-866-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR