2,6-Pyridindimethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,6-Bis(hydroxymethyl)pyridine

M-Clarity™ Qualitätsstufe = MQ100

154369-100G 154369-25G 154369-5G
SIAL154369-100GEA 429 EUR
SIAL154369-100G SIAL154369-25G SIAL154369-5G
2,6-Pyridindimethanol, Sigma-Aldrich®
2,6-Pyridindimethanol
Formel: C₇H₉NO₂
Molecular Weight: 139.15 g/mol
MDL: MFCD00006351
CAS-Nummer: 1195-59-1
EINECS: 214-803-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR