3-Pyridinboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Pyridine-3-boronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

15449-1G
SIAL15449-1GEA 40.4 EUR
SIAL15449-1G
3-Pyridinboronsäurepinakolester, Sigma-Aldrich®
3-Pyridinboronsäurepinakolester
Formel: C₁₁H₁₆BNO₂
Molecular Weight: 205,06 g/mol
MDL: MFCD03084758
CAS-Nummer: 329214-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR