1-Boc-piperazin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate

15502-25G 15502-5G
SIAL15502-25GEA 250 EUR
SIAL15502-25G SIAL15502-5G
1-Boc-piperazin, Sigma-Aldrich®
1-Boc-piperazin
Formel: C₉H₁₈N₂O₂
MDL: MFCD00075265
CAS-Nummer: 57260-71-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR