1-Boc-piperazin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate
15502-25G
15502-5G
SIAL15502-25GEA
250
EUR
SIAL15502-25G
SIAL15502-5G
1-Boc-piperazin, Sigma-Aldrich®
1-Boc-piperazin
Formel:
C₉H₁₈N₂O₂ |
MDL:
MFCD00075265 CAS-Nummer: 57260-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...