N(α)-Boc-L-tryptophan 99,0 TLC_ASSAY_METHOD, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Boc-Trp-OH

15512-5G-F
SIAL15512-5G-FEA 76.4 EUR
SIAL15512-5G-F
N(α)-Boc-L-tryptophan 99,0 TLC_ASSAY_METHOD, Sigma-Aldrich®
N(α)-Boc-L-tryptophan
Formel: C₁₆H₂₀N₂O₄
Molecular Weight: 304.3 g/mol
MDL: MFCD00065595
CAS-Nummer: 13139-14-5
EINECS: 236-072-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR