Methyl-3-nitrobenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Nitrobenzoesäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

155977-100G
SIAL155977-100GEA 63.5 EUR
SIAL155977-100G
Methyl-3-nitrobenzoat, Sigma-Aldrich®
Methyl-3-nitrobenzoat
Formel: C₈H₇NO₄
Molecular Weight: 181,15 g/mol
MDL: MFCD00007250
CAS-Nummer: 618-95-1
EINECS: 210-574-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR