(R)-(-)-3-Aminopiperidin Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-(-)-3-Piperidinamine dihydrochloride, (R)-Piperidin-3-amine dihydrochloride

15626-1G-F
SIAL15626-1G-FEA 70.4 EUR
SIAL15626-1G-F
(R)-(-)-3-Aminopiperidin Dihydrochlorid, Sigma-Aldrich®
(R)-(-)-3-Aminopiperidin Dihydrochlorid
Formel: C₅H₁₄Cl₂N₂
Molecular Weight: 173,08 g/mol
MDL: MFCD06799458
CAS-Nummer: 334618-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR