Cyclopropyltriphenylphosphoniumbromid, Sigma-Aldrich®

Lieferant: Merck

157317-25G
SIAL157317-25GEA 312 EUR
SIAL157317-25G
Cyclopropyltriphenylphosphoniumbromid, Sigma-Aldrich®
Cyclopropyltriphenylphosphonium bromide
Formel: C₂₁H₂₀BrP
Molecular Weight: 383,27 g/mol
Schmelzpunkt: 189…190 °C
MDL: MFCD00011872
CAS-Nummer: 14114-05-7
EINECS: 237-970-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR