3,5-Dichloranisol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,3-Dichloro-5-methoxybenzene

M-Clarity™ Qualitätsstufe = MQ200

157341-25G
SIAL157341-25GEA 97.9 EUR
SIAL157341-25G
3,5-Dichloranisol, Sigma-Aldrich®
3,5-Dichloranisol
Formel: C₇H₆Cl₂O
MDL: MFCD00000589
CAS-Nummer: 33719-74-3
EINECS: 251-655-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR