3,5-Di-tert-butyl-o-benzochinon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

157457-10G
SIAL157457-10GEA 217 EUR
SIAL157457-10G
3,5-Di-tert-butyl-o-benzochinon, Sigma-Aldrich®
3,5-Di-tert-butyl-o-benzochinon
Formel: C₁₄H₂₀O₂
MDL: MFCD00001647
CAS-Nummer: 3383-21-9
EINECS: 222-189-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR