4-Nitrochalkon, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Nitrobenzylidenacetophenone
M-Clarity™ Qualitätsstufe = MQ100
157481-25G
SIAL157481-25GEA
380
EUR
SIAL157481-25G
4-Nitrochalkon, Sigma-Aldrich®
4-Nitrochalkon
Formel:
C₁₅H₁₁NO₃ Molecular Weight: 253,26 g/mol Schmelzpunkt: 165 °C |
MDL:
MFCD00007382 CAS-Nummer: 1222-98-6 EINECS: 214-949-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...