4-Brom-1,3-dimethoxybenzol, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
1-Bromo-2,4-dimethoxybenzene
157554-25G
SIAL157554-25GEA
33.6
EUR
SIAL157554-25G
4-Brom-1,3-dimethoxybenzol, Sigma-Aldrich®
4-Brom-1,3-dimethoxybenzol
Formel:
C₈H₉BrO₂ Molecular Weight: 217,06 g/mol Siedepunkt: 282 °C (1013 hPa) Dichte: 1,507 g/cm³ (20 °C) |
MDL:
MFCD00009844 CAS-Nummer: 17715-69-4 EINECS: 241-717-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...