2-Chloroethanesulphonic acid sodium salt hydrate ≥98%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Sodium-2-chloroethanesulphonate hydrate

M-Clarity™ Qualitätsstufe = MQ200

157651-25G
SIAL157651-25GEA 125 EUR
SIAL157651-25G
2-Chloroethanesulphonic acid sodium salt hydrate ≥98%, Sigma-Aldrich®
2-Chloroethanesulphonic acid sodium salt hydrate
Formel: C₂H₄ClNaO₃S.H₂O
Molecular Weight: 184,57 g/mol
MDL: MFCD00007531
CAS-Nummer: 15484-44-3
EINECS: 241-935-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR