1-Phenylcyclopropancarbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

160075-10G
SIAL160075-10GEA 85 EUR
SIAL160075-10G
1-Phenylcyclopropancarbonsäure, Sigma-Aldrich®
1-Phenylcyclopropanecarboxylic acid
Formel: C₁₀H₁₀O₂
MDL: MFCD00001288
CAS-Nummer: 6120-95-2
EINECS: 228-090-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR