3-Amino-1H-pyrazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Pyrazolamine, 3-AP, 1H-Pyrazol-3-amine, 3-Aminopyrazole

M-Clarity™ Qualitätsstufe = MQ100

160644-10G 160644-2.5G
SIAL160644-10GEA 237 EUR
SIAL160644-10G SIAL160644-2.5G
3-Amino-1H-pyrazol, Sigma-Aldrich®
3-Amino-1H-pyrazole
Formel: C₃H₅N₃
Molecular Weight: 83.09 g/mol
MDL: MFCD00005236
CAS-Nummer: 1820-80-0
EINECS: 217-338-1
UN: 3259
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR