Tris(6,6,7,7,8,8,8-heptafluor-2,2-dimethyl-3,5-octandionato-O,O')europium(III) 99, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Europium(III)-tris(1,1,1,2,2,3,3-heptafluoro-7,7-dimethyl-4,6-octanedionate), Eu(fod)3, Resolve-Al™ EuFOD, Europium-fod, Siever's reagent

160938-1G 160938-5G
SIAL160938-1GEA 75.3 EUR
SIAL160938-1G SIAL160938-5G
Tris(6,6,7,7,8,8,8-heptafluor-2,2-dimethyl-3,5-octandionato-O,O')europium(III) 99, Sigma-Aldrich®
Tris(6,6,7,7,8,8,8-heptafluor-2,2-dimethyl-3,5-octandionato-O,O')europium(III)
Formel: C₃₀H₃₀EuF₂₁O₆
Molecular Weight: 1037.49 g/mol
MDL: MFCD00064655
CAS-Nummer: 17631-68-4
EINECS: 241-616-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR