5-(4-Hydroxyphenyl)-5-phenylhydantoin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

161543-1G
SIAL161543-1GEA 126 EUR
SIAL161543-1G
5-(4-Hydroxyphenyl)-5-phenylhydantoin, Sigma-Aldrich®
5-(4-Hydroxyphenyl)-5-phenylhydantoin
Formel: C₁₅H₁₂N₂O₃
CAS-Nummer: 2784-27-2
EINECS: 220-492-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR