Bis(4-nitrophenyl)carbonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Bis(4-nitrophenyl) carbonate

M-Clarity™ Qualitätsstufe = MQ200

161691-10G
SIAL161691-10GEA 114 EUR
SIAL161691-10G
Bis(4-nitrophenyl)carbonat, Sigma-Aldrich®
Bis(4-nitrophenyl)carbonat
Formel: C₁₃H₈N₂O₇
Molecular Weight: 304,22 g/mol
MDL: MFCD00007322
CAS-Nummer: 5070-13-3
EINECS: 225-775-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR