Di(2-pyridyl)ketonoxim, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2'-Dipyridyl ketoxime

161705-1G
SIAL161705-1GEA 69.2 EUR
SIAL161705-1G
Di(2-pyridyl)ketonoxim, Sigma-Aldrich®
Di(2-pyridyl)ketonoxim
Formel: C₁₁H₉N₃O
Molecular Weight: 199,21 g/mol
MDL: MFCD00006311
CAS-Nummer: 1562-95-4
EINECS: 216-349-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR