1,3-Diethyl-2-thiobarbitursäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,3-Diethyl-2-thioxodihydropyrimidine-4,6(1H,5H)-dione

M-Clarity™ Qualitätsstufe = MQ100

161950-50G
SIAL161950-50GEA 392 EUR
SIAL161950-50G
1,3-Diethyl-2-thiobarbitursäure, Sigma-Aldrich®
1,3-Diethyl-2-thiobarbitursäure
Formel: C₈H₁₂N₂O₂S
Molecular Weight: 200,260 g/mol
MDL: MFCD00006676
CAS-Nummer: 5217-47-0
EINECS: 226-010-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR