4-Hydroxy-3-methoxybenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

162604-25G
SIAL162604-25GEA 56.2 EUR
SIAL162604-25G
4-Hydroxy-3-methoxybenzonitril, Sigma-Aldrich®
4-Hydroxy-3-methoxybenzonitril
Formel: C₈H₇NO₂
Molecular Weight: 149,15 g/mol
MDL: MFCD00001820
CAS-Nummer: 4421-08-3
EINECS: 224-589-0
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR