2-Picolylchlorid Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

162701-25G 162701-100G 162701-5G
SIAL162701-25GEA 88.7 EUR
SIAL162701-25G SIAL162701-100G SIAL162701-5G
2-Picolylchlorid Hydrochlorid, Sigma-Aldrich®
2-Picolylchlorid Hydrochlorid
Formel: C₆H₇Cl₂N
MDL: MFCD00012811
CAS-Nummer: 6959-47-3
EINECS: 230-149-9
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR