α-Brom-4-nitro-o-kresol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(Brommethyl)-4-nitrophenol, 2-Hydroxy-5-nitrobenzylbromid, Koshland Reagenz I
162906-5G
SIAL162906-5GEA
59.7
EUR
SIAL162906-5G
α-Brom-4-nitro-o-kresol, Sigma-Aldrich®
α-Brom-4-nitro-o-kresol
Formel:
C₇H₆BrNO₃ Molecular Weight: 232,03 g/mol Schmelzpunkt: 147 °C Dichte: >1 g/cm³ (20 °C) |
MDL:
MFCD00007340 CAS-Nummer: 772-33-8 EINECS: 212-248-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...