2-(2-(2-Chlorethoxy)ethoxy)ethanol, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

162973-10G
SIAL162973-10GEA 70 EUR
SIAL162973-10G
2-(2-(2-Chlorethoxy)ethoxy)ethanol, Sigma-Aldrich®
2-(2-(2-Chloroethoxy)ethoxy)ethanol
Formel: C₆H₁₃ClO₃
Molecular Weight: 168,62 g/mol
MDL: MFCD00002874
CAS-Nummer: 5197-62-6
EINECS: 225-984-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR