2-Methyl-1-tetralon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methyl-3,4-dihydronaphthalen-1(2H)-one

M-Clarity™ Qualitätsstufe = MQ100

163228-2.5G
SIAL163228-2.5GEA 132 EUR
SIAL163228-2.5G
2-Methyl-1-tetralon, Sigma-Aldrich®
2-Methyl-1-tetralon
Formel: C₁₁H₁₂O
Molecular Weight: 160,22 g/mol
MDL: MFCD00001690
CAS-Nummer: 1590-08-5
EINECS: 216-461-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR